Alfa Aesar™ Nickel powder, APS 3-7 micron, 99.9% (metals basis)

Manufacturer: Alfa Aesar™ 1025609

Quantity 10g

 View more versions of this product

Catalog No. AA1025609

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7440-02-0
    MDL Number MFCD00011137
    UN Number UN3089
    Chemical Name or Material Nickel powder

    Specifications

    Health Hazard 3 GHS P Statement
    P210-P260-P261-P363-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wash contaminated clothing before reuse.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Particle Size APS 3 to 7 Micron
    Health Hazard 2 GHS H Statement
    H372-H351-H228-H317
    Causes damage to organs through prolonged or repeated exposure.
    Suspected of causing cancer.
    Flammable solid.
    May cause an allergic skin reaction.
    Quantity 10g
    MDL Number MFCD00011137
    UN Number UN3089
    Health Hazard 1 Danger
    TSCA TSCA
    Physical Form Powder
    CAS 7440-02-0
    Odor Odorless
    DOT Information Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: METAL POWDERS, FLAMMABLE, N.O.S.
    RTECS Number QR5950000
    Weight Apparent density: 1.8g/cm3 to 2.7g/cm3
    Chemical Name or Material Nickel powder
    Assay 99.9%, (metals basis)

    GHS H Statement
    H372-H351-H228-H317
    Causes damage to organs through prolonged or repeated exposure.
    Suspected of causing cancer.
    Flammable solid.
    May cause an allergic skin reaction.

    GHS P Statement
    P210-P260-P261-P363-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wash contaminated clothing before reuse.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit