Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Nickel powder, spherical, -100+325 mesh, 99.8% (metals basis)

Manufacturer:  Alfa Aesar™ 42733A1

Quantity 1kg

 View more versions of this product

Catalog No. AA42733A1


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

7440-02-0
MFCD00011137
UN3089
Nickel powder

Specifications

1kg
Sperical Powder
UN3089
Odorless
Nickel powder
7440-02-0
MFCD00011137
−100+325 Mesh
Gray
99.8%, (metals basis)

Safety and Handling

SDS & Safety Documents

GHS H Statement
H372-H351-H228-H317
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing cancer.
Flammable solid.
May cause an allergic skin reaction.

GHS P Statement
P210-P260-P261-P363-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wash contaminated clothing before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: METAL POWDERS, FLAMMABLE, N.O.S.

RTECSNumber : QR5950000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit