Alfa Aesar™ Nickel powder, spherical, -100+325 mesh, 99.8% (metals basis)

Manufacturer: Alfa Aesar™ 4273318

Quantity 50g

 View more versions of this product

Catalog No. AA4273318

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7440-02-0
    MDL Number MFCD00011137
    UN Number UN3089
    Chemical Name or Material Nickel powder

    Specifications

    Physical Form Sperical Powder
    Health Hazard 3 GHS P Statement
    P210-P260-P261-P363-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wash contaminated clothing before reuse.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00011137
    UN Number UN3089
    Odor Odorless
    TSCA TSCA
    Mesh Size −100+325 Mesh
    Quantity 50g
    CAS 7440-02-0
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H372-H351-H228-H317
    Causes damage to organs through prolonged or repeated exposure.
    Suspected of causing cancer.
    Flammable solid.
    May cause an allergic skin reaction.
    Color Gray
    DOT Information Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: METAL POWDERS, FLAMMABLE, N.O.S.
    RTECS Number QR5950000
    Chemical Name or Material Nickel powder
    Assay 99.8%, (metals basis)

    GHS H Statement
    H372-H351-H228-H317
    Causes damage to organs through prolonged or repeated exposure.
    Suspected of causing cancer.
    Flammable solid.
    May cause an allergic skin reaction.

    GHS P Statement
    P210-P260-P261-P363-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wash contaminated clothing before reuse.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit