Nickel Powder, Spherical, -300 mesh, 99.8% (Metals basis), Alfa Aesar™

Manufacturer: Alfa Aesar 1058122

Quantity 100g

 View more versions of this product

Catalog No. AA1058122

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7440-02-0
    Physical Form Spherical Powder
    Assay Percent Range 99.8% (Metals basis)
    MDL Number MFCD00011137
    Chemical Name or Material Nickel

    Specifications

    Physical Form Spherical Powder
    Quantity 100g
    Color Silver-gray
    DOT Information DOT Class: 4.2, Packaging Group: II
    Health Hazard 2 Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). Avoid release to the environment. Refer to special instructions/Safety data sheets.
    EINECS Number 231-111-4
    Assay Percent Range 99.8% (Metals basis)
    MDL Number MFCD00011137
    CAS 7440-02-0
    Odor Odorless
    Health Hazard 1 H372-H351-H228-H317 : Causes damage to organs through prolonged or repeated exposure. Suspected of causing cancer. Flammable solid. May cause an allergic skin reaction
    Appearance Silver gray powder of flakes
    Chemical Name or Material Nickel

    Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). Avoid release to the environment. Refer to special instructions/Safety data sheets.

    H372-H351-H228-H317 : Causes damage to organs through prolonged or repeated exposure. Suspected of causing cancer. Flammable solid. May cause an allergic skin reaction

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit