Alfa Aesar™ Nickel powder, spherical, -325 mesh (5% max. +325), 99.7% min. (metals basis)b

Manufacturer: Alfa Aesar™ 77725P2

Quantity 1 lb.

 View more versions of this product

Catalog No. AA77725P2

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7440-02-0
    MDL Number MFCD00011137
    UN Number UN3089
    Chemical Name or Material Nickel

    Specifications

    Physical Form Spherical Powder
    Health Hazard 3 GHS P Statement
    P210-P222-P363-P405-P422a-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Do not allow contact with air.
    Wash contaminated clothing before reuse.
    Store locked up.
    Store contents under inert gas.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00011137
    Health Hazard 2 GHS H Statement
    H228-H250-H351-H317
    Flammable solid.
    Catches fire spontaneously if exposed to air.
    Suspected of causing cancer.
    May cause an allergic skin reaction.
    TSCA TSCA
    UN Number UN3089
    Mesh Size −325 mesh (5% max. +325)
    Odor Odorless
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 4.1; Packaging Group: II; Proper Shipping Name: METAL POWDERS, FLAMMABLE, N.O.S.
    RTECS Number QR5950000
    Quantity 1 lb.
    CAS 7440-02-0
    Chemical Name or Material Nickel
    Assay ≥99.7%, (metals basis)

    GHS H Statement
    H228-H250-H351-H317
    Flammable solid.
    Catches fire spontaneously if exposed to air.
    Suspected of causing cancer.
    May cause an allergic skin reaction.

    GHS P Statement
    P210-P222-P363-P405-P422a-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Do not allow contact with air.
    Wash contaminated clothing before reuse.
    Store locked up.
    Store contents under inert gas.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit