Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nickel, Reference Standard Solution (1000ppm ±1%/Certified), Fisher Chemical

Click to view available options
Quantity:
100mL
500mL
Packaging:
Poly Bottle
13478-00-7,7697-37-2,7732-18-5
H12N2NiO12
290.791
MFCD00011137
AOPCKOPZYFFEDA-UHFFFAOYSA-N
61630
nickel(2+);dinitrate;hexahydrate
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.[Ni+2]
13478-00-7,7697-37-2,7732-18-5
H12N2NiO12
290.791
MFCD00011137
AOPCKOPZYFFEDA-UHFFFAOYSA-N
61630
nickel(2+);dinitrate;hexahydrate
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.[Ni+2]
Concentration 990 to 1010ppm
Concentration or Composition (by Analyte or Components) 1000ppm ±1%
Physical Form Liquid
Grade Certified
Packaging Poly Bottle
Traceability to NIST LOT NUMBER USED NICKEL REF. STANDARD
Vapor Pressure 14mmHg at 20°C
Boiling Point 100°C
Color Blue Green
Quantity 500mL
Chemical Name or Material Nickel, Reference Standard Solution, 1000ppm ±1%
Show More Show Less

DOTInformation : DOT Class 8, : Corrosive