Alfa Aesar™ Nickel silicide, 99% (metals basis excluding Co), Co 0.1-1%

Manufacturer: Alfa Aesar™ 3625130

Quantity 250g

 View more versions of this product

Catalog No. AA3625130

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 12059-14-2
    Formula Weight 145.47
    MDL Number MFCD00151367
    UN Number UN3077
    Chemical Name or Material Nickel silicide

    Specifications

    Formula Weight 145.47
    Health Hazard 3 GHS P Statement
    P260-P261-P280-P363-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Wash contaminated clothing before reuse.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Odor Odorless
    Physical Form Powder
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H350-H372-H317
    May cause cancer.
    Causes damage to organs through prolonged or repeated exposure.
    May cause an allergic skin reaction.
    TSCA TSCA
    Mesh Size −20 Mesh
    UN Number UN3077
    DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
    MDL Number MFCD00151367
    CAS 12059-14-2
    Quantity 250g
    Chemical Name or Material Nickel silicide
    Assay 99%, (metals basis excluding Co), Co 0.1 to 1%

    GHS H Statement
    H350-H372-H317
    May cause cancer.
    Causes damage to organs through prolonged or repeated exposure.
    May cause an allergic skin reaction.

    GHS P Statement
    P260-P261-P280-P363-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Wash contaminated clothing before reuse.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit