Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nickel Sponge, 2% Iron and 2.5% Chromium, Alfa Aesar™

Manufacturer:  Alfa Aesar 4486622

Quantity 100g

 View more versions of this product

Catalog No. AA4486622


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

Sponge, 2% Iron and 2.5% Chromium
MFCD03844839
Nickel

Specifications

100g
Sponge, 2% Iron and 2.5% Chromium
Dark gray
Dark gray sponge
MFCD03844839
Nickel

Safety and Handling

SDS & Safety Documents

Wear suitable protective clothing and gloves.

H251-H351-H317 : Self-heating: may catch fire. Suspected of causing cancer. May cause an allergic skin reaction

DOTInformation : DOT Class: 4.2, Packaging Group: II

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit