Nickel Sponge, 2% Iron and 2.5% Chromium, Alfa Aesar™

Manufacturer: Alfa Aesar 4486636

Quantity 500g

 View more versions of this product

Catalog No. AA4486636

  Check Availability
Add to Cart

    Chemical Identifiers

    Physical Form Sponge, 2% Iron and 2.5% Chromium
    MDL Number MFCD03844839
    Chemical Name or Material Nickel

    Specifications

    Quantity 500g
    Color Dark gray
    DOT Information DOT Class: 4.2, Packaging Group: II
    Physical Form Sponge, 2% Iron and 2.5% Chromium
    MDL Number MFCD03844839
    Health Hazard 1 H251-H351-H317 : Self-heating: may catch fire. Suspected of causing cancer. May cause an allergic skin reaction
    Appearance Dark gray sponge
    Health Hazard 2 Wear suitable protective clothing and gloves.
    Chemical Name or Material Nickel

    Wear suitable protective clothing and gloves.

    H251-H351-H317 : Self-heating: may catch fire. Suspected of causing cancer. May cause an allergic skin reaction

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit