Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nickel Standard, 1000mg/L in 2% HNO3, (Assurance Grade), SPEX CertiPrep™

Catalog Number p-219084
Click to view available options
Quantity:
30 mL
250 mL
Packaging:
Poly Bottle
Triple-leached LDPE bottle
7440-02-0, 7697-37-2, 7732-18-5
Ni
58.69
MFCD00011137 MFCD06798735
PXHVJJICTQNCMI-UHFFFAOYSA-N
935
CHEBI:28112
nickel
[Ni]
7440-02-0, 7697-37-2, 7732-18-5
Ni
58.69
MFCD00011137 MFCD06798735
PXHVJJICTQNCMI-UHFFFAOYSA-N
935
CHEBI:28112
nickel
[Ni]

Includes

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Ni1000μg/mL

Certifications

ISO/IEC 17025:2005, ISO 17034:2016, ISO 9001: 2015

Compliance

GHS/OSHA Complaint

Packaging Triple-leached LDPE bottle
Quantity 30 mL
CAS Min % 0.1, 2.0, 97.9
CAS Max % 0.1, 2.0, 97.9
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Grade Assurance
Concentration or Composition (by Analyte or Components) Matrix    Concentration    Cat. No.
2-5% HNO3    1000μg/mL    PLNI2-2Y, -2T, -2X    10,000    PLNI2-3Y, -3X
Solvent or Matrix 2% HNO3
Chemical Name or Material Nickel Standard
Hazard Category Causes skin irritation, serious eye irritation. May cause respiratory irritation
DOTInformation Nonhazardous
Recommended Storage Ambient
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit