Learn More
Nickel standard solution, for AAS, 1 mg/ml Ni in 2% HNO3, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 196155000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For atomic adsorption spectroscopySpecifications
Nickel standard solution | |
1 mg/ml Ni in 2% HNO3 | |
Plastic bottle | |
Green | |
2.1, 97.1 | |
MFCD00011137 MFCD06798735 | |
[Ni] | |
58.69 | |
CHEBI:28112 | |
AAS | |
1.032 | |
Crystals, Crystalline Powder and/or Chunks |
990 to 1010μg/mL Ni in 2 to 5% HNO3 (20°C) | |
7440-02-0, 13478-00-7, 7697-37-2, 7732-18-5 | |
500 mL | |
1.9, 96.9 | |
Ni | |
PXHVJJICTQNCMI-UHFFFAOYSA-N | |
nickel | |
935 | |
58.69 | |
1.0320g/mL | |
Soluble in water |
Safety and Handling
GHS H Statement
May cause cancer by inhalation. May damage the unborn child. Causes severe skin burns and eye damage. May produce an allergic reaction. Harmful to aquatic life with long lasting effects. May be corrosive to metals.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Keep only in original container.
Danger
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
RUO – Research Use Only