Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nickel sulfide, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog Number AA3566118
Click to view available options
Quantity:
10 g
50 g
250 g
12035-72-2
Ni3S2
240.20
MFCD01711880
YGHCWPXPAHSSNA-UHFFFAOYSA-N
nickel subsulfide, trinickel disulphide, nickel sulfide, trinickel disulfide, nickel subsulphide, nickel sesquisulfide, nickel tritadisulphide, nickel iii sulfide, alpha nickel subsulfide, ccris 432
22833473
S=[Ni][Ni][Ni]=S
12035-72-2
Ni3S2
240.20
MFCD01711880
YGHCWPXPAHSSNA-UHFFFAOYSA-N
nickel subsulfide, trinickel disulphide, nickel sulfide, trinickel disulfide, nickel subsulphide, nickel sesquisulfide, nickel tritadisulphide, nickel iii sulfide, alpha nickel subsulfide, ccris 432
22833473
S=[Ni][Ni][Ni]=S

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 797°C
Color Black
Physical Form Powder
Quantity 50 g
Assay Percent Range 99.9% (metals basis)
Solubility Information Soluble in water(0,00735g/L at 20°C).
IUPAC Name nickel;(sulfanylidene-$l^{4}-sulfanylidene)nickel
Formula Weight 240.19
Appearance Black powder
Chemical Name or Material Nickel sulfide
Hazard Category H317-H341-H350i-H372
Precautionary Statement P201-P202-P260-P264b-P270-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c
DOTInformation DOT Class: 9; Packing Group: III
EINECSNumber 234-829-6
RTECSNumber QR9800000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit