Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nickel sulfide, 99.9% (metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 3566130

Quantity 250g

 View more versions of this product

Catalog No. AA3566130


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

12035-72-2
99.9% (metals basis)
240.19
Nickel sulfide
Powder
Ni3S2
MFCD00016266

Specifications

12035-72-2
99.9% (metals basis)
Black powder
MFCD00016266
250g
Powder
Ni3S2
240.19
Black
Nickel sulfide

Safety and Handling

SDS & Safety Documents

H350-H372-H341-H400-H410-H317: May cause cancer. Causes damage to organs through prolonged or repeated exposure. Suspected of causing genetic defects. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects. May cause an allergic skin reaction.

DOTInformation : DOT Class: 9; Packing Group: III

EINECSNumber : 234-829-6

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit