Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Nickel Thinfoil, 0.0025mm (0.0001 in.) thick, 99.95% (metals basis)

Click to view available options
Quantity:
25 x 25mm
50 x 50mm
100 x 100mm
7440-02-0
Ni
58.693
MFCD00011137
PXHVJJICTQNCMI-UHFFFAOYSA-N
raney alloy,fibrex,catalyst,particles,fibrex p,nickel, elemental,nichel italian,nickel, soluble salts,carbonyl powder,niccolum
935
CHEBI:28112
nickel
[Ni]
7440-02-0
Ni
58.693
MFCD00011137
PXHVJJICTQNCMI-UHFFFAOYSA-N
raney alloy,fibrex,catalyst,particles,fibrex p,nickel, elemental,nichel italian,nickel, soluble salts,carbonyl powder,niccolum
935
CHEBI:28112
nickel
[Ni]
Quantity 25 x 25mm
Weight ≈0.014g/25x25mm
Odor Odorless
Physical Form Foil
Percent Purity 99.95%
Chemical Name or Material Nickel Thinfoil, 0.0025mm (0.0001 in.) thick, Not certified pinhole free
Assay (metals basis)

GHS H Statement
H372-H351-H317
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing cancer.
May cause an allergic skin reaction.

P201-P202-P260-P261-P264b-P270-P272-P280-P302+P352-P308+P313-P333+P313-P363-P501c

H317-H351-H372

EINECSNumber : 231-111-4

TSCA : Yes

Recommended Storage : Ambient temperatures