Nickel Thinfoil, 0.006mm (0.0002 in.) thick, hard, 99.95% (Metals basis), Alfa Aesar™

Temper: as rolled; Form: Approx.4.0g/25 x 30cm, not certified pinhole free

Manufacturer: Alfa Aesar 14368-FI

Quantity 50 x 50mm

 View more versions of this product

Catalog No. AA14368FI

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 7440-02-0
    Physical Form Thin Foil, 0.006mm (0.0002 in.) thick
    Assay Percent Range 99.95% (Metals basis)
    MDL Number MFCD00011137
    Chemical Name or Material Nickel, Hard

    Specifications

    Physical Form Thin Foil, 0.006mm (0.0002 in.) thick
    Quantity 50 x 50mm
    Color Silver-gray
    EINECS Number 231-111-4
    Temper Temper: as rolled
    Assay Percent Range 99.95% (Metals basis)
    MDL Number MFCD00011137
    CAS 7440-02-0
    Odor Odorless
    Health Hazard 1 H372-H351-H317 : Causes damage to organs through prolonged or repeated exposure. Suspected of causing cancer. May cause an allergic skin reaction
    Appearance Silver gray Thinfoil
    Health Hazard 2 Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).
    Chemical Name or Material Nickel, Hard

    Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).

    H372-H351-H317 : Causes damage to organs through prolonged or repeated exposure. Suspected of causing cancer. May cause an allergic skin reaction

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit