Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nickel Thinfoil, 0.006mm (0.0002 in.) thick, hard, 99.95% (Metals basis), Alfa Aesar™

Temper: as rolled; Form: Approx.4.0g/25 x 30cm, not certified pinhole free

Manufacturer:  Alfa Aesar 14368GH

Quantity 100 x 100mm

 View more versions of this product

Catalog No. AA14368GH

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Chemical Identifiers

7440-02-0
99.95% (Metals basis)
Nickel, Hard
Thin Foil, 0.006mm (0.0002 in.) thick
MFCD00011137

Specifications

100 x 100mm
Silver gray Thinfoil
Thin Foil, 0.006mm (0.0002 in.) thick
MFCD00011137
Temper: as rolled
Nickel, Hard
7440-02-0
8.908g/cm3 at 20°C
99.95% (Metals basis)
Odorless
Silver-gray

Safety and Handling

Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).

H372-H351-H317 : Causes damage to organs through prolonged or repeated exposure. Suspected of causing cancer. May cause an allergic skin reaction

EINECSNumber : 231-111-4

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit