Learn More
Alfa Aesar™ Nickel Ultrathin foil, 0.25 micron thick, 99.95% (metals basis)
Manufacturer:  Alfa Aesar™ 40712FF
Quantity 25x25mm
 View more versions of this product
Catalog No. AA40712FF
  Check Availability
Add to cart
Specifications Description & Specifications
Chemical Identifiers
7440-02-0
MFCD00011137
Nickel Ultrathin foil
Specifications
25x25mm
On Temporary Acrylic Support
Odorless
Nickel Ultrathin foil
7440-02-0
MFCD00011137
0.25 Micron thick
99.95%, (metals basis)
Safety and Handling
SDS & Safety Documents
GHS H Statement
H372-H351-H317
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing cancer.
May cause an allergic skin reaction.
GHS P Statement
P260-P261-P280-P363-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash contaminated clothing before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
TSCA : TSCA