Alfa Aesar™ Nickel Vanadium, sputtering target, 76.2mm (3.0 in.) dia. x 3.18mm (0.125 in.) thick, 99.95% (metals basis)

Manufacturer: Alfa Aesar™ 40880KS

Quantity 1 each

 View more versions of this product

Catalog No. AA40880KS

  Check Availability
Add to Cart

    Chemical Identifiers

    MDL Number MFCD01310520
    Chemical Name or Material Nickel Vanadium, sputtering target, 76.2mm (3.0 in.) dia. x 3.18mm (0.125 in.) thick

    Specifications

    Physical Form Non-Magnetic
    MDL Number MFCD01310520
    Avg. Mol. Wt. or Mol. Wt. Range Ni:V; 93:7 wt%
    Quantity 1 each
    Chemical Name or Material Nickel Vanadium, sputtering target, 76.2mm (3.0 in.) dia. x 3.18mm (0.125 in.) thick
    Assay 99.95%, (metals basis)

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit