Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nickel Wire, 0.5mm (0.02 in.) dia., 99.5% (Metals basis), Alfa Aesar™

Form: Approx.1.75g/m, Temper: soft

Manufacturer:  Alfa Aesar 10250H9

Quantity 500m

 View more versions of this product

Catalog No. AA10250H9


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

7440-02-0
99.5% (Metals basis)
Nickel
Wire, 0.5mm (0.02 in.) dia.
MFCD00011137

Specifications

500m
Silver gray wire
99.5% (Metals basis)
Odorless
Nickel
7440-02-0
Wire, 0.5mm (0.02 in.) dia.
MFCD00011137
Silver-gray

Safety and Handling

SDS & Safety Documents

Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).

H372-H351-H317 : Causes damage to organs through prolonged or repeated exposure. Suspected of causing cancer. May cause an allergic skin reaction

EINECSNumber : 231-111-4

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit