Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nickelous Carbonate, Powder, BAKER ANALYZED™ Reagent, J.T.Baker™

Click to view available options
Quantity:
1.5kg
Packaging:
Poly Bottle

Chemical Identifiers

3333-67-3
CNiO3
118.701
ZULUUIKRFGGGTL-UHFFFAOYSA-L
18746

High quality chemicals for laboratory and specialized industrial use

CAS 3333-67-3
Molecular Formula CNiO3
InChI Key ZULUUIKRFGGGTL-UHFFFAOYSA-L
SMILES C(=O)([O-])[O-].[Ni+2]
IUPAC Name nickel(2+);carbonate
Molecular Weight (g/mol) 118.701
PubChem CID 18746
Formula Weight 118.7
Physical Form Powder
Packaging Poly Bottle
Quantity 1.5kg
Chemical Name or Material Nickelous Carbonate
Grade BAKER ANALYZED™ Reagent
Show More Show Less