Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nickelous Chloride, 6-Hydrate, Crystal, BAKER ANALYZED™ Reagent, J.T.Baker™

Click to view available options
Quantity:
125g
500g
Packaging:
Poly Bottle

Chemical Identifiers

7791-20-0
NiCl2·6H2O

High quality chemicals for laboratory and specialized industrial use

Formula Weight 237.71
Physical Form Crystals
Packaging Poly Bottle
Specific Gravity 1L = 3.55kg
Quantity 500g
Chemical Name or Material Nickelous Chloride, 6-Hydrate
Grade BAKER ANALYZED™ Reagent