Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nickelous Sulfate, 6-Hydrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
125g
500g
Packaging:
Poly Bottle

Chemical Identifiers

10101-97-0
H12NiO10S
262.839
RRIWRJBSCGCBID-UHFFFAOYSA-L
5284429
CHEBI:53437

High quality chemicals for laboratory and specialized industrial use

SMILES O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[Ni+2]
IUPAC Name nickel(2+);sulfate;hexahydrate
Formula Weight 262.85
Physical Form Crystals
Packaging Poly Bottle
Specific Gravity 1L = 2.03kg
Quantity 125g
Chemical Name or Material Nickelous Sulfate, 6-Hydrate
Grade Baker Analyzed™ ACS Reagent