Alfa Aesar™ Nicofuranose

Manufacturer: Alfa Aesar™ B2202436

Quantity 500g

 View more versions of this product

Catalog No. AAB2202436

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 15351-13-0
    Molecular Formula C30H24N4O10
    Formula Weight 600.52
    MDL Number MFCD00868276
    Synonym beta-D-Fructofuranose tetranicotinate
    Chemical Name or Material Nicofuranose

    Specifications

    Formula Weight 600.52
    Molecular Formula C30H24N4O10
    MDL Number MFCD00868276
    CAS 15351-13-0
    Synonym beta-D-Fructofuranose tetranicotinate
    Odor Odorless
    Melting Point 133° to 136°C
    Quantity 500g
    Chemical Name or Material Nicofuranose

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit