Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
β-Nicotinamide adenine dinucleotide phosphate, disodium salt, 98-100%, MP Biomedicals™
Click to view available options
Quantity:
50 mg
100 mg
250 mg
500 mg
1 g
5 g
Specifications
Color | White |
Assay Percent Range | 98-100% |
Quantity | 50 mg |
Formula Weight | 787.4 |
Physical Form | Powder |
Chemical Name or Material | β-Nicotinamide adenine dinucleotide phosphate, disodium salt and monopotassium salt |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction