Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

β-Nicotinamide Adenine Dinucleotide Phosphate, Monosodium Salt, Tetrahydrate, Affymetrix/USB™

Click to view available options
Quantity:
250mg
500mg
1g
5g
Packaging:
Plastic bottle
1184-16-3
C21H28N7NaO17P3
766.398
MFCD00036973
ZUXQIVQXILFYQI-QYZPTAICSA-N
nadp sodium salt,beta-nicotinamide adenine dinucleotide phosphate sodium
131845262
[[(2R,3R,4R,5R)-5-(6-aminopurin-9-yl)-3-hydroxy-4-phosphonooxyoxolan-2-yl]methoxy-hydroxyphosphoryl] [(2R,3S,4R,5R)-5-(3-carbamoylpyridin-1-ium-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphate;sodium
C1=CC(=C[N+](=C1)C2C(C(C(O2)COP(=O)([O-])OP(=O)(O)OCC3C(C(C(O3)N4C=NC5=C4N=CN=C5N)OP(=O)(O)O)O)O)O)C(=O)N.[Na]
1184-16-3
C21H28N7NaO17P3
766.398
MFCD00036973
ZUXQIVQXILFYQI-QYZPTAICSA-N
nadp sodium salt,beta-nicotinamide adenine dinucleotide phosphate sodium
131845262
[[(2R,3R,4R,5R)-5-(6-aminopurin-9-yl)-3-hydroxy-4-phosphonooxyoxolan-2-yl]methoxy-hydroxyphosphoryl] [(2R,3S,4R,5R)-5-(3-carbamoylpyridin-1-ium-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphate;sodium
C1=CC(=C[N+](=C1)C2C(C(C(O2)COP(=O)([O-])OP(=O)(O)OCC3C(C(C(O3)N4C=NC5=C4N=CN=C5N)OP(=O)(O)O)O)O)O)C(=O)N.[Na]
Formula Weight 837.45
Physical Form Powder
Packaging Plastic bottle
Quantity 1g
Chemical Name or Material beta-Nicotinamide adenine dinucleotide phosphate monosodium salt tetrahydrate