Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nicotinamide N-Methyltransferase/NNMT Mouse anti-Human, Alexa Fluor™ 750, Clone: AT11G11, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250581AF750
Description
Nicotinamide N-Methyltransferase/NNMT Monoclonal specifically detects Nicotinamide N-Methyltransferase/NNMT in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
Nicotinamide N-Methyltransferase/NNMT | |
Monoclonal | |
Alexa Fluor 750 | |
50 mM sodium borate with 0.05% sodium azide | |
NNMT | |
Recombinant human NNMT (1-264aa) purified from E. coli | |
100 μL | |
4837.0 | |
Store at 4°C in the dark. |
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
AT11G11 | |
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence | |
EC 2.1.1.1, nicotinamide N-methyltransferase | |
Mouse | |
Protein A purified | |
Primary | |
Human | |
IgG2a κ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title