Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nicotinamide Test Substance, For Elemental Analysis, MilliporeSigma™

Click to view available options
Quantity:
5g
98-92-0
C6H6N2O
122.127
DFPAKSUCGFBDDF-UHFFFAOYSA-N
nicotinamide, niacinamide, 3-pyridinecarboxamide, vitamin pp, nicotinic acid amide, papulex, aminicotin, amixicotyn, nicobion, nicotylamide
936
CHEBI:17154
pyridine-3-carboxamide
C1=CC(=CN=C1)C(=O)N
98-92-0
C6H6N2O
122.127
DFPAKSUCGFBDDF-UHFFFAOYSA-N
nicotinamide, niacinamide, 3-pyridinecarboxamide, vitamin pp, nicotinic acid amide, papulex, aminicotin, amixicotyn, nicobion, nicotylamide
936
CHEBI:17154
pyridine-3-carboxamide
C1=CC(=CN=C1)C(=O)N
Ignition Point 480°C
Packaging Glass bottle
Quantity 5g
Physical Form Crystals
Chemical Name or Material Nicotinamide

H319: Causes serious eye irritation.