Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(±)-Nicotine-d3 salicylate, Honeywell Fluka™

Click to view available options
Quantity:
10 mg
1173021-00-5
1173021-00-5

Analytical standard, reference material

Ensure you have the information you need to quickly start your analysis, and pass relevant audits:

  • High quality inorganic and organic reference materials produced according to ISO 9001
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results
  • Free expert technical support
Physical Form Solid
Grade Analytical Standard
Packaging Glass Bottle
Quantity 10 mg
For Use With (Application) ALKALOIDS