Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nicotinic Acetylcholine R alpha 1/CHRNA1, Rat anti-Human,Mouse,Rat, Clone: mAb 192, Novus Biologicals™

Rat Monoclonal Antibody

$135.50 - $419.00

Specifications

Antigen Nicotinic Acetylcholine R alpha 1/CHRNA1
Clone mAb 192
Applications Western Blot, ELISA, Blocking Assay
Host Species Rat
Target Species Human, Mouse, Rat
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB015339
View Documents
Novus Biologicals
NBP2810700.025MG
0.025 mg
Each for $135.50
Add to cart
 
NB015340
View Documents
Novus Biologicals
NBP2810700.2MG
0.2 mg
Each for $419.00
Add to cart
 
Description

Description

Nicotinic Acetylcholine R alpha 1/CHRNA1 Monoclonal antibody specifically detects Nicotinic Acetylcholine R alpha 1/CHRNA1 in Human, Mouse, Rat Samples. It is validated for Western Blot, ELISA, Blocking Assay.
Specifications

Specifications

Nicotinic Acetylcholine R alpha 1/CHRNA1
Western Blot, ELISA, Blocking Assay
Human, Mouse, Rat
Primary
mAb 192
Rat
Monoclonal
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit