Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nicotinic Acetylcholine R alpha 9/CHRNA9 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Specifications
Antigen | Nicotinic Acetylcholine R alpha 9/CHRNA9 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB023236
|
Novus Biologicals
NBP294341100UL |
100 μg | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
NB023237
|
Novus Biologicals
NBP29434120UL |
20 μg | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Description
Nicotinic Acetylcholine R alpha 9/CHRNA9 Polyclonal antibody specifically detects Nicotinic Acetylcholine R alpha 9/CHRNA9 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
Nicotinic Acetylcholine R alpha 9/CHRNA9 | |
Polyclonal | |
Purified | |
RUO | |
PBS (pH 7.3), 50% glycerol | |
55584 | |
IgG | |
Affinity purified |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
cholinergic receptor, nicotinic, alpha 9, cholinergic receptor, nicotinic, alpha polypeptide 9, HSA243342, MGC142109, MGC142135, NACHR alpha 9, NACHR alpha-9, NACHRA9acetylcholine receptor, neuronal nicotinic, alpha-9 subunit, neuronal acetylcholine receptor protein, alpha-9 subunit, neuronal acetylcholine receptor subunit alpha-9, nicotinic acetylcholine receptor subunit alpha 9, Nicotinic acetylcholine receptor subunit alpha-9 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 26-240 of human CHRNA9 (NP_060051.2). | |
Primary | |
Store at -20°:C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title