Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nicotinic acid, 99%, Thermo Scientific Chemicals

Catalog Number aaa1268330
Click to view available options
Quantity:
250 g
1000 g
5000 g
59-67-6
C6H5NO2
123.11
MFCD00006391
PVNIIMVLHYAWGP-UHFFFAOYSA-N
nicotinic acid, niacin, 3-pyridinecarboxylic acid, 3-carboxypyridine, wampocap, acidum nicotinicum, apelagrin, pellagrin, akotin, daskil
938
CHEBI:15940
pyridine-3-carboxylic acid
OC(=O)C1=CC=CN=C1
59-67-6
C6H5NO2
123.11
MFCD00006391
PVNIIMVLHYAWGP-UHFFFAOYSA-N
nicotinic acid, niacin, 3-pyridinecarboxylic acid, 3-carboxypyridine, wampocap, acidum nicotinicum, apelagrin, pellagrin, akotin, daskil
938
CHEBI:15940
pyridine-3-carboxylic acid
OC(=O)C1=CC=CN=C1

Vitamin of the B complex with hypolipidemic properties occurring in various animal and plant tissues. Required by the body for the formation of coenzymes NAD and NADP.Nicotinic acid is used as an antidyslipidemic agent. It is also used to improve all lipoproteins well above the vitamin requirement and lowers the total cholesterol, bad LDL-cholesterol, and triglyceride levels. Further, it is used as a food additive in food industries, cosmetics and pharmaceuticals. In addition to this, it is used in medication, nutrient, enriched flours and in electroplating baths.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water. Slightly soluble in ethanol and ethyl ether. Insoluble in most lipid solvents.

Notes
Light sensitive. Store in a cool place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 236°C to 239°C
Density 1.4
Flash Point 293°C (559°F)
Assay Percent Range 99%
Quantity 250 g
Beilstein 109591
Merck Index 14,6525
Solubility Information Soluble in water. Slightly soluble in ethanol and ethyl ether. Insoluble in most lipid solvents.
Formula Weight 123.11
Percent Purity 99%
Chemical Name or Material Nicotinic Acid
Show More Show Less
Hazard Category H319
Hazard Statement GHS H Statement
H319
Precautionary Statement P264b-P280i-P305+P351+P338
EINECSNumber 200-441-0
RTECSNumber QT0525000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit