Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nicotinic Acid Hydrazide 97%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Plastic bottle
553-53-7
C6H7N3O
137.14
MFCD00006383
KFUSANSHCADHNJ-UHFFFAOYSA-N
nicotinohydrazide,nicotinic acid hydrazide,nicotinic hydrazide,niazid,nicotinylhydrazide,nicotinoyl hydrazine,3-pyridoyl hydrazine,3-pyridylcarbonylhydrazine,nicotinoyl hydrazide,hydrazine, nicotinoyl
11112
pyridine-3-carbohydrazide
C1=CC(=CN=C1)C(=O)NN
553-53-7
C6H7N3O
137.14
MFCD00006383
KFUSANSHCADHNJ-UHFFFAOYSA-N
nicotinohydrazide,nicotinic acid hydrazide,nicotinic hydrazide,niazid,nicotinylhydrazide,nicotinoyl hydrazine,3-pyridoyl hydrazine,3-pyridylcarbonylhydrazine,nicotinoyl hydrazide,hydrazine, nicotinoyl
11112
pyridine-3-carbohydrazide
C1=CC(=CN=C1)C(=O)NN
Packaging Plastic bottle
Melting Point 160°C to 163°C
Quantity 100g
Infrared Spectrum Authentic
Color White
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Beilstein 22, 41
Merck Index 09, 6345
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol,slightly soluble in benzene
Formula Weight 137.14
Physical Form Fine Needle-Like Crystalline Powder
Percent Purity ≥96.0%
Chemical Name or Material Nicotinic acid hydrazide, 97%
Show More Show Less

GHS H Statement:
Suspected of causing cancer.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Use personal protective equipment as required.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 209-041-0

RTECSNumber : QT1750000