Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Nicotinuric acid, 98+%

Click to view available options
Quantity:
5g
25g
583-08-4
C8H8N2O3
180.163
MFCD00023578
ZBSGKPYXQINNGF-UHFFFAOYSA-N
nicotinuric acid,nicotinylglycine,nicotinoylglycine,2-nicotinamido acetic acid,n-nicotinoylglycine,n-pyridin-3-ylcarbonyl glycine,nicotinoyl-glycine,n-nicotinylglycine,nicotinurate,n-nicotinoyl-glycine
68499
CHEBI:7563
2-(pyridine-3-carbonylamino)acetic acid
C1=CC(=CN=C1)C(=O)NCC(=O)O
583-08-4
C8H8N2O3
180.163
MFCD00023578
ZBSGKPYXQINNGF-UHFFFAOYSA-N
nicotinuric acid,nicotinylglycine,nicotinoylglycine,2-nicotinamido acetic acid,n-nicotinoylglycine,n-pyridin-3-ylcarbonyl glycine,nicotinoyl-glycine,n-nicotinylglycine,nicotinurate,n-nicotinoyl-glycine
68499
CHEBI:7563
2-(pyridine-3-carbonylamino)acetic acid
C1=CC(=CN=C1)C(=O)NCC(=O)O
Applications
Measurement of NUA levels in urine is used to indicate possible elevated nicotinamide adenine dinucleotide (NAD) catabolism. NUA may be used as a reference (standard curve) in analysis of NAD and NUA catabolism. It is an antiproliferative.

Solubility
Soluble in dimethyl sulfoxide, methanol, water, and 1M NH4OH (50 mg/mL).

Notes
Store at -20°C. Incompatible with oxidizing agents.
Color White
Quantity 5g
Formula Weight 180.16
Physical Form Crystalline Powder
Percent Purity ≥98%
Chemical Name or Material Nicotinuric acid