Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nidogen-1/Entactin Mouse anti-Human, Alexa Fluor™ 532, Clone: 2R33, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP222037AF532

Catalog No. NP222037A32


Add to cart

Description

Description

Nidogen-1/Entactin Monoclonal antibody specifically detects Nidogen-1/Entactin in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin).
Specifications

Specifications

Nidogen-1/Entactin
2R33
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Conjugated, Purified
NID1
This antibody was produced from a hybridoma (mouse myeloma fused with spleen cells from a mouse immunized with purified recombinant Nidogen-1/Entactin. Mouse anti human Nidogen-1/Entacin NP_002499.
Protein G purified
Store at 4°C in the dark.
Monoclonal
Human
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Alexa Fluor™ 532
50 mM sodium borate with 0.05% sodium azide
enactin, Entactin, NID-1, NIDentactin, nidogen (enactin), nidogen 1, nidogen-1
Mouse
IgG1
0.1 mL
Primary
4811.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit