Learn More
Nidogen-1/Entactin Mouse anti-Human, Alexa Fluor™ 750, Clone: 2R33, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP222037AF750
Catalog No. NP222037A70
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Nidogen-1/Entactin Monoclonal antibody specifically detects Nidogen-1/Entactin in Human samples. It is validated for Western Blot,ELISA,Immunohistochemistry,Immunohistochemistry (Paraffin).
Specifications
Specifications
Nidogen-1/Entactin
2R33
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
enactin, Entactin, NID-1, NIDentactin, nidogen (enactin), nidogen 1, nidogen-1
Mouse
IgG1
Store at 4°C in the dark.
Monoclonal
Human
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Alexa Fluor™ 750
50 mM sodium borate with 0.05% sodium azide
NID1
This antibody was produced from a hybridoma (mouse myeloma fused with spleen cells from a mouse immunized with purified recombinant Nidogen-1/Entactin. Mouse anti human Nidogen-1/Entacin NP_002499.
0.1 mL
Primary
4811