Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nidogen-1/Entactin Rat anti-Mouse, Guinea Pig, Clone: ELM1, Novus Biologicals™

Rat Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP30751510UG

 View more versions of this product

Catalog No. NB122503


Add to cart

Description

Description

Nidogen-1/Entactin Monoclonal specifically detects Nidogen-1/Entactin in Mouse, Guinea Pig samples. It is validated for ELISA.
Specifications

Specifications

Nidogen-1/Entactin
ELM1
Unconjugated
10 mM PBS with 0.05% BSA
enactin, Entactin, NID-1, NIDentactin, nidogen (enactin), nidogen 1, nidogen-1
Murine EHS laminin preparation
Protein A or G purified
RUO
Primary
4811
ELISA
0.2 mg/ml
ELISA 2-4 ug/ml
Purified
Rat
IgG2a Kappa
100 μg
Store at 4°C.
Monoclonal
Mouse, Guinea Pig
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit