Learn More
Niedner™ Delta Hydro Fire Hose Tester
Unique check valve system closes automatically in the event of hose failure without shutting down the other lines
Manufacturer:  NIEDNER CDHT2000SS
 View more versions of this product
Catalog No. 19-816-110
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
2 in. Stainless Steel manifold, 1.7hp motor, 2.5 in Aluminium NST check valves, Stainless Steel cart, 110/120V, NST thread, 4.3 gpm Oil bath pump, 2.5 in. NST female swivel inlet connector, Inline filter
500psi
50.34kg
Stainless Steel
111 lb.
Fire hose tester