Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Niemann-Pick type C2 Antibody Pair [HRP]
Rabbit Monoclonal Antibody Pair
Supplier: Novus Biologicals™ NBP27941315PLATES
Description
This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human Niemann-Pick type C2.Specifications
156.25 pg/mL | |
Sandwich ELISA | |
Antibody Pairs | |
Human | |
EDDM1, epididymal secretory protein E1, He1, HE1epididymal protein 1, Human epididymis-specific protein 1, MGC1333, Niemann-Pick disease type C2 protein, Niemann-Pick disease, type C2, NP-C2, tissue-specific secretory protein | |
NPC2 | |
RUO | |
Niemann-Pick type C2 |
156.25-10000 pg/mL | |
HRP | |
This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human Niemann-Pick type C2. | |
Sandwich ELISA | |
10577 | |
Rabbit | |
15 Plates | |
Storage is content dependent. | |
1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in PBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.15 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 10000 pg/mL is recommended. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction