Learn More
ABclonal Technology Niemann-Pick type C2NPC2
SDP
Supplier:  ABclonal Technology A23128100UL
Encompass_Preferred
This gene encodes a protein containing a lipid recognition domain The encoded protein may function in regulating the transport of cholesterol through the late endosomal lysosomal system Mutations in this gene have been associated with Niemann-Pick disease type C2 and frontal lobe atrophy
Catalog No. 50-000-06036
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.