Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NIF3L1 Mouse anti-Canine, Human, Mouse, Rat, Clone: OTI5A7, liquid, TrueMAB
Mouse Monoclonal Antibody
Manufacturer: OriGene TA503680
Description
ALS2CR1 may function as a transcriptional corepressor through its interaction with COPS2, negatively regulating the expression of genes involved in neuronal differentiation.Specifications
NIF3L1 | |
Monoclonal | |
0.66 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q4V7D6, Q9EQ80, Q9GZT8 | |
ALS2CR1, CALS-7, MDS015 | |
Mouse | |
IgG2a | |
100 μL | |
-20° C, Avoid Freeze/Thaw Cycles | |
Primary | |
301431, 60491, 607446, 65102 |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
OTI5A7 | |
Unconjugated | |
NIF3L1 | |
Liquid | |
NIF3L1 | |
Full length human recombit protein of human NIF3L1 produced in HEK293T cell. | |
Affinity Chromatography | |
RUO | |
Antibody | |
Monoclonal | |
Canine, Human, Mouse, Rat |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title