Alfa Aesar™ Nigrosin, soluble in spirit

Manufacturer: Alfa Aesar™ B2142414

Quantity 25g

 View more versions of this product

Catalog No. AAB2142414

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 11099-03-9
    MDL Number MFCD00149005
    Synonym C.I. 50415; Solvent Black 5
    Chemical Name or Material Nigrosin

    Specifications

    Melting Point >300°C
    TSCA TSCA
    MDL Number MFCD00149005
    CAS 11099-03-9
    Synonym C.I. 50415; Solvent Black 5
    Solubility Information Soluble in spirit
    RTECS Number GE5800000
    Quantity 25g
    Chemical Name or Material Nigrosin

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit