Learn More
Nigrosin water soluble, Thermo Scientific Chemicals
Description
It is employed as a common biological stain for tissues and cells. Used to make inks and shoe-polish and for dyeing leather, wood, textiles, and plastics. The crude mixture of nigrosine hydrochlorides (Phenol Black L) is used to dye phenolic resins. Bases are used to make printing colors and to dye plastics. It finds its use in Dorner?s spore stain, as a negative stain in place of India ink for spirochetes, bacteria, protozoa and fungi. It is also used in compounding resins include phenolic resins, styrenics, polyamides and urea resins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
It is employed as a common biological stain for tissues and cells. Used to make inks and shoe-polish and for dyeing leather, wood, textiles, and plastics. The crude mixture of nigrosine hydrochlorides (Phenol Black L) is used to dye phenolic resins. Bases are used to make printing colors and to dye plastics. It finds its use in Dorner′s spore stain, as a negative stain in place of India ink for spirochetes, bacteria, protozoa and fungi. It is also used in compounding resins include phenolic resins, styrenics, polyamides and urea resins.
Solubility
Soluble in water, ethanol, oil, nigrosines and fat.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
Quantity | 100 g |
Chemical Name or Material | Nigrosin, water soluble |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.