Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nimesulide, Tocris Bioscience™ Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
100mg
This item is not returnable. View return policy
51803-78-2
C13H12N2O5S
308.308
HYWYRSMBCFDLJT-UHFFFAOYSA-N
nimesulide,mesulid,n-4-nitro-2-phenoxyphenyl methanesulfonamide,flogovital,sulidene,nimed,4-nitro-2-phenoxymethanesulfonanilide,nisulid,nimesulidum inn-latin,nimesulida inn-spanish
4495
CHEBI:44445
N-(4-nitro-2-phenoxyphenyl)methanesulfonamide
CS(=O)(=O)NC1=C(C=C(C=C1)[N+](=O)[O-])OC2=CC=CC=C2
This item is not returnable. View return policy
51803-78-2
C13H12N2O5S
308.308
HYWYRSMBCFDLJT-UHFFFAOYSA-N
nimesulide,mesulid,n-4-nitro-2-phenoxyphenyl methanesulfonamide,flogovital,sulidene,nimed,4-nitro-2-phenoxymethanesulfonanilide,nisulid,nimesulidum inn-latin,nimesulida inn-spanish
4495
CHEBI:44445
N-(4-nitro-2-phenoxyphenyl)methanesulfonamide
CS(=O)(=O)NC1=C(C=C(C=C1)[N+](=O)[O-])OC2=CC=CC=C2

Cyclooxygenase-2 (COX-2) inhibitor

Selective, orally active cyclooxygenase-2 (COX-2) inhibitor. Produces potent analgesic, anti-inflammatory and antipyretic activities in vivo. Reported to produce fewer gastrointestinal side effects than standard NSAIDs.??.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Quantity 100mg
Formula Weight 308.31
Percent Purity >99%
Chemical Name or Material Nimesulide

Recommended Storage : Store at +4°C