Learn More
Ninhydrin, 99%, Thermo Scientific Chemicals
Description
Ninhydrin is used for the determination of amino groups in amino acids, peptides and proteins. It finds application in forensic science for the analysis of latent fingerprints on porous surfaces such as paper. It is also used in thin layer chromatography (TLC) to monitor the progress of chemical reactions. In Kaiser test, it plays an important role to monitor the deprotection in solid phase peptide synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Ninhydrin is used for the determination of amino groups in amino acids, peptides and proteins. It finds application in forensic science for the analysis of latent fingerprints on porous surfaces such as paper. It is also used in thin layer chromatography (TLC) to monitor the progress of chemical reactions. In Kaiser test, it plays an important role to monitor the deprotection in solid phase peptide synthesis.
Solubility
Soluble in water, acetone, ethanol, and methanol. Slightly soluble in chloroform and ether.
Notes
Light Sensitive. Incompatible with strong oxidizing agents, amines, alkalis and strong bases.
Specifications
Melting Point | 251°C (decomposition) |
Assay Percent Range | 99% |
Quantity | 10 g |
Beilstein | 1910963 |
Sensitivity | Light sensitive |
Merck Index | 14,6554 |
Solubility Information | Soluble in water,acetone,ethanol,and methanol. Slightly soluble in chloroform and ether. |
Formula Weight | 178.14 |
Percent Purity | 99% |
Physical Form | Powder |
Show More |
Safety and Handling
Hazard Category | H302-H315-H319-H335 |
Hazard Statement | GHS H Statement H302-H315-H319-H335 |
Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c |
EINECSNumber | 207-618-1 |
RTECSNumber | NK5425000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.