Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ninhydrin, Monohydrate, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

missing translation for 'orderingAttributeHoverText'
Quantity:
10g
25g
Packaging:
Wide Mouth Amber Glass Bottle
485-47-2
C9H6O4
178.143
FEMOMIGRRWSMCU-UHFFFAOYSA-N
ninhydrin,ninhydrin hydrate,1,2,3-indantrione monohydrate,ninhydrine,2,2-dihydroxy-1h-indene-1,3 2h-dione,2,2-dihydroxy-1,3-indandione,triketohydrindene hydrate,1h-indene-1,3 2h-dione, 2,2-dihydroxy,1,2,3-indantrione, 2-hydrate,2,2-dihydroxyindane-1,3-dione
10236
CHEBI:86374
2,2-dihydroxyindene-1,3-dione
C1=CC=C2C(=C1)C(=O)C(C2=O)(O)O
485-47-2
C9H6O4
178.143
FEMOMIGRRWSMCU-UHFFFAOYSA-N
ninhydrin,ninhydrin hydrate,1,2,3-indantrione monohydrate,ninhydrine,2,2-dihydroxy-1h-indene-1,3 2h-dione,2,2-dihydroxy-1,3-indandione,triketohydrindene hydrate,1h-indene-1,3 2h-dione, 2,2-dihydroxy,1,2,3-indantrione, 2-hydrate,2,2-dihydroxyindane-1,3-dione
10236
CHEBI:86374
2,2-dihydroxyindene-1,3-dione
C1=CC=C2C(=C1)C(=O)C(C2=O)(O)O

High quality chemicals for laboratory and specialized industrial use

Packaging Wide Mouth Amber Glass Bottle
Quantity 25g
Formula Weight 178.14
Grade BAKER ANALYZED™ ACS Reagent
Chemical Name or Material Ninhydrin, Monohydrate