Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ninhydrin, Monohydrate, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
10g
25g
Packaging:
Wide Mouth Amber Glass Bottle

Chemical Identifiers

485-47-2
C9H6O4
178.143
FEMOMIGRRWSMCU-UHFFFAOYSA-N
10236
CHEBI:86374

High quality chemicals for laboratory and specialized industrial use

Quantity 10g
CAS 485-47-2
Packaging Wide Mouth Amber Glass Bottle
Molecular Formula C9H6O4
InChI Key FEMOMIGRRWSMCU-UHFFFAOYSA-N
SMILES C1=CC=C2C(=C1)C(=O)C(C2=O)(O)O
IUPAC Name 2,2-dihydroxyindene-1,3-dione
Molecular Weight (g/mol) 178.143
PubChem CID 10236
ChEBI CHEBI:86374
Formula Weight 178.14
Chemical Name or Material Ninhydrin, Monohydrate
Grade Baker Analyzed™ ACS Reagent
Show More Show Less