Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Nintedanib

Potent VEGFR, PDGFR and FGFR inhibitor

$62.00 - $261.50

Specifications

CAS 656247-17-5
Chemical Name or Material Methyl (Z)-3-[[[4-(N-methyl-2-(4-methylpiperazin-1-yl)acetamido)phenyl)amino)(phenyl)methylene]-2-oxoindoline-6-carboxylate
Molecular Formula C31H33N5O4
Purity 98%
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
70-491-0
View Documents
Tocris Bioscience™
7049/10
10 mg
Each for $62.00
Add to cart
 
70-495-0
View Documents
Tocris Bioscience™
7049/50
50 mg
Each for $261.50
Add to cart
 
Description

Description

  • Potent inhibitor of vascular endothelial growth factor receptors (VEGFRs), platelet-derived growth factor receptors (PDGFRs) and fibroblast growth factor receptors (FGFRs) (IC50 values are 13 to 34 nM for VEGFR subtypes, 59 to 65 nM for PDGFR subtypes, 37 to 108 nM for FGFR subtypes).
  • Also inhibits Flt-3, Lck, Lyn and Src (IC50 values are 25 to 200 nM).
  • Inhibits proliferation in VEGF-stimulated endothelial cells, and PDGF-stimulated pericytes and vascular smooth muscle cells.
  • Reduces vessel density and integrity, and inhibits tumor growth in a mouse tumor xenograft model.
Specifications

Specifications

656247-17-5
Methyl (Z)-3-[[[4-(N-methyl-2-(4-methylpiperazin-1-yl)acetamido)phenyl)amino)(phenyl)methylene]-2-oxoindoline-6-carboxylate
C31H33N5O4
98%
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit