Niobium Foil, 0.025 ± 0.015mm (0.001 ± 0.0006in) thick, 0.998, Alfa Aesar™

Form: Approx.8.6g/20 x 20cm

Manufacturer: Alfa Aesar 10263CV

Quantity 20 x 100cm

 View more versions of this product

Catalog No. AA10263CV

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    CAS 7440-03-1
    Physical Form Foil, 0.025 ± 0.015mm (0.001 ± 0.0006 in.) thick
    Assay Percent Range 99.80%
    MDL Number MFCD00011126
    Chemical Name or Material Niobium

    Specifications

    Quantity 20 x 100cm
    Color Silver-gray
    Physical Form Foil, 0.025 ± 0.015mm (0.001 ± 0.0006 in.) thick
    Assay Percent Range 99.80%
    MDL Number MFCD00011126
    CAS 7440-03-1
    Odor Odorless
    Appearance Silver gray foil
    Chemical Name or Material Niobium

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit