Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Niobium Foil, 0.025 ± 0.015mm (0.001 ± 0.0006in) thick, 0.998, Alfa Aesar™

Form: Approx.8.6g/20 x 20cm

Manufacturer:  Alfa Aesar 10263CU

Quantity 20 x 20cm

 View more versions of this product

Catalog No. AA10263CU


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

7440-03-1
99.80%
Niobium
Foil, 0.025 ± 0.015mm (0.001 ± 0.0006 in.) thick
MFCD00011126

Specifications

20 x 20cm
Silver gray foil
99.80%
Odorless
Niobium
7440-03-1
Foil, 0.025 ± 0.015mm (0.001 ± 0.0006 in.) thick
MFCD00011126
Silver-gray
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit