Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Niobium gauze, 40 mesh woven from 0.178mm (0.007 in.) dia. wire

Click to view available options
Quantity:
75 x 75mm
150 x 150mm
300 x 300mm
7440-03-1
Nb
92.91
MFCD00011126
GUCVJGMIXFAOAE-UHFFFAOYSA-N
columbium,element,niobium-93,powder,foil,rod,vn 1,foil, 0.25mm 0.01in thick,columbio,niobio
23936
CHEBI:33344
niobium
[Nb]
7440-03-1
Nb
92.91
MFCD00011126
GUCVJGMIXFAOAE-UHFFFAOYSA-N
columbium,element,niobium-93,powder,foil,rod,vn 1,foil, 0.25mm 0.01in thick,columbio,niobio
23936
CHEBI:33344
niobium
[Nb]

Filtration, electrodes

Quantity 300 x 300mm
Mesh Size 40 Mesh
Physical Form Wire Cloth
Chemical Name or Material Niobium gauze, Woven from 0.178mm (0.007 in.) dia. wire

EINECSNumber : 231-113-5

TSCA : Yes

Recommended Storage : Ambient temperatures